Assay Details
Target Gene Details
Entrez Gene ID: | 830 |
Gene Name: | capping actin protein of muscle Z-line alpha subunit 2 |
Gene Aliases: |
CAPPA2, CAPZ |
Location: |
Chr.7:116862478-116919261 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZA2 | NM_006136.2 | NP_006127.1 | ||
| AK222989.1 | BAD96709.1 | |||
| AK223004.1 | BAD96724.1 | |||
| AK293405.1 | ||||
| AK294409.1 | ||||
| AK314808.1 | ||||
| AK315915.1 | ||||
| BC005338.1 | AAH05338.1 | |||
| BI553815.1 | ||||
| BT006735.1 | AAP35381.1 | |||
| DQ655955.2 | ||||
| DQ655956.2 | ||||
| U03269.1 | AAA88848.1 | |||
| U03851.1 | AAC00534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831107 | Chr.7:116774680 - 117009412 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map