Assay Details
Target Gene Details
Entrez Gene ID: | 101928268 |
Gene Name: | uncharacterized LOC101928268 |
Gene Aliases: |
- |
Location: |
Chr.7:30460050-30469217 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928268 | XM_005249925.3 | XP_005249982.1 |
Target Gene Details
Entrez Gene ID: | 10392 |
Gene Name: | nucleotide binding oligomerization domain containing 1 |
Gene Aliases: |
CARD4, CLR7.1, NLRC1 |
Location: |
Chr.7:30424527-30478840 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map