Assay Details
Target Gene Details
Entrez Gene ID: | 285954 |
Gene Name: | INHBA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:41693916-41779378 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INHBA-AS1 | NR_027118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2671244 | Chr.7:41719014 - 41719777 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114609819] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map