Assay Details
Target Gene Details
Entrez Gene ID: | 5898 |
Gene Name: | RALA Ras like proto-oncogene A |
Gene Aliases: |
RAL |
Location: |
Chr.7:39622955-39708124 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALA | NM_005402.3 | NP_005393.2 | ||
| XM_006715762.2 | XP_006715825.1 | |||
| XM_011515466.1 | XP_011513768.1 | |||
| AK308109.1 | ||||
| AU132355.1 | ||||
| BC039858.1 | AAH39858.1 | |||
| DC313757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508451 | Chr.7:39621185 - 39747623 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117467174,rs78234728] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map