Assay Details
Target Gene Details
Entrez Gene ID: | 401409 |
Gene Name: | RAB19, member RAS oncogene family |
Gene Aliases: |
RAB19B |
Location: |
Chr.7:140403740-140426881 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB19 | XM_005249996.3 | 4 | 1718 | XP_005250053.1 |
| XM_005249997.3 | 4 | 1701 | XP_005250054.1 | |
| XM_005249998.3 | 4 | 1696 | XP_005250055.1 | |
| XM_017012207.1 | 4 | 1631 | XP_016867696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608500 | Chr.7:140408296 - 140517870 on Build GRCh38 | Loss |
|
| nsv522190 | Chr.7:140366611 - 140430863 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map