Assay Details
Target Gene Details
Entrez Gene ID: | 4232 |
Gene Name: | mesoderm specific transcript |
Gene Aliases: |
PEG1 |
Location: |
Chr.7:130486175-130506297 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEST | NM_001253900.1 | NP_001240829.1 | ||
| NM_001253901.1 | NP_001240830.1 | |||
| NM_001253902.1 | NP_001240831.1 | |||
| NM_002402.3 | NP_002393.2 | |||
| NM_177524.2 | NP_803490.1 | |||
| NM_177525.2 | NP_803491.1 | |||
| XM_011516222.1 | XP_011514524.1 | |||
| AK098397.1 | ||||
| BC002413.2 | AAH02413.1 | |||
| BC011908.2 | AAH11908.1 | |||
| BC014564.2 | AAH14564.1 | |||
| BC018695.2 | AAH18695.1 | |||
| BC090049.1 | AAH90049.1 | |||
| BG831799.1 | ||||
| BI459412.1 | ||||
| CR627364.1 | CAH10465.1 | |||
| D78611.1 | BAA11432.1 | |||
| D87367.1 | BAA21757.1 | |||
| DA487705.1 | ||||
| DB454206.1 | ||||
| DC367784.1 | ||||
| Y11534.1 | CAA72297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831131 | Chr.7:130468986 - 130656327 on Build GRCh38 | Loss |
|
| nsv831130 | Chr.7:130418592 - 130510992 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map