Assay Details
Target Gene Details
Entrez Gene ID: | 84182 |
Gene Name: | family with sequence similarity 188 member B |
Gene Aliases: |
C7orf67 |
Location: |
Chr.7:30771417-30892387 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM188B | NM_032222.2 | NP_115598.2 | ||
| AF538962.1 | AAQ10898.1 | |||
| AK026027.1 | BAB15327.1 | |||
| BC098569.1 | AAH98569.1 | |||
| DA868176.1 |
Target Gene Details
Entrez Gene ID: | 100526825 |
Gene Name: | INMT-FAM188B readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.7:30752135-30892387 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INMT-FAM188B | NR_037598.1 | |||
| AK308762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606507 | Chr.7:30755672 - 30801825 on Build GRCh38 | Loss |
|
| dgv6316n100 | Chr.7:30609596 - 31145482 on Build GRCh38 | Gain |
|
| nsv830938 | Chr.7:30644614 - 30857907 on Build GRCh38 | Gain |
|
| nsv527038 | Chr.7:30617851 - 31122331 on Build GRCh38 | Gain |
|
| esv2761316 | Chr.7:30615466 - 31127062 on Build GRCh38 | Gain |
|
| nsv606508 | Chr.7:30772130 - 30797387 on Build GRCh38 | Loss |
|
| nsv830939 | Chr.7:30704855 - 30871891 on Build GRCh38 | Loss |
|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78059384] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map