Assay Details
Target Gene Details
Entrez Gene ID: | 3624 |
Gene Name: | inhibin beta A subunit |
Gene Aliases: |
EDF, FRP |
Location: |
Chr.7:41685101-41710532 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INHBA | XM_017012174.1 | 5 | 5166 | XP_016867663.1 |
| XM_017012175.1 | 3 | 4850 | XP_016867664.1 | |
| XM_017012176.1 | 5 | 5034 | XP_016867665.1 |
More Information
Set Membership: |
|

Genomic Map