Assay Details
Target Gene Details
Entrez Gene ID: | 669 |
Gene Name: | bisphosphoglycerate mutase |
Gene Aliases: |
DPGM |
Location: |
Chr.7:134646779-134679816 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPGM | NM_001293085.1 | NP_001280014.1 | ||
| NM_001724.4 | NP_001715.1 | |||
| NM_199186.2 | NP_954655.1 | |||
| AK097778.1 | ||||
| AK315439.1 | ||||
| BC017050.1 | AAH17050.1 | |||
| BM090687.1 | ||||
| BU664387.1 | ||||
| BX473275.1 | ||||
| CB994648.1 | ||||
| CF242942.1 | ||||
| DA701935.1 | ||||
| DB047822.1 | ||||
| DB049554.1 | ||||
| X04327.1 | CAA27858.1 |
More Information
Set Membership: |
|

Genomic Map