Assay Details
Target Gene Details
Entrez Gene ID: | 10049 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B6 |
Gene Aliases: |
DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1 |
Location: |
Chr.7:157337004-157417439 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB6 | NM_005494.2 | NP_005485.1 | ||
| NM_058246.3 | NP_490647.1 | |||
| XM_005249515.2 | XP_005249572.1 | |||
| XM_005249516.2 | XP_005249573.1 | |||
| XM_006715823.1 | XP_006715886.1 | |||
| XM_011515704.1 | XP_011514006.1 | |||
| XM_017011633.1 | XP_016867122.1 | |||
| AB014888.1 | BAA32209.1 | |||
| AB015798.1 | BAA88769.1 | |||
| AB015799.1 | BAA88770.1 | |||
| AB209859.1 | BAD93096.1 | |||
| AF060703.1 | AAF21257.1 | |||
| AF075601.1 | AAD43194.1 | |||
| AK223601.1 | BAD97321.1 | |||
| AK291953.1 | ||||
| AK293025.1 | ||||
| AK297497.1 | ||||
| AK297796.1 | ||||
| AK297989.1 | ||||
| AK301152.1 | ||||
| AL136707.1 | CAB66642.1 | |||
| BC000177.2 | AAH00177.1 | |||
| BC002446.2 | AAH02446.1 | |||
| EU075177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3615688 | Chr.7:157288516 - 157403484 on Build GRCh38 | Gain |
|
| esv2735675 | Chr.7:157288269 - 157403297 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map