Assay Details
Target Gene Details
Entrez Gene ID: | 11335 |
Gene Name: | chromobox 3 |
Gene Aliases: |
HECH, HP1-GAMMA, HP1Hs-gamma |
Location: |
Chr.7:26201211-26213607 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBX3 | NM_007276.4 | NP_009207.2 | ||
| NM_016587.3 | NP_057671.2 | |||
| DB565723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv970454 | Chr.7:26211886 - 26214604 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map