Assay Details
Target Gene Details
Entrez Gene ID: | 57001 |
Gene Name: | succinate dehydrogenase complex assembly factor 3 |
Gene Aliases: |
ACN9, DC11, LYRM10, Sdh7 |
Location: |
Chr.7:97116593-97181763 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHAF3 | NM_020186.2 | NP_064571.1 | ||
| AF201933.1 | AAF86869.1 | |||
| BC020621.1 | AAH20621.1 | |||
| BC022858.1 | AAH22858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614222 | Chr.7:97100233 - 97121364 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map