Assay Details
Target Gene Details
Entrez Gene ID: | 113263 |
Gene Name: | glucocorticoid induced 1 |
Gene Aliases: |
FAM117C, GCTR, GIG18, TSSN1 |
Location: |
Chr.7:7968743-8089080 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLCCI1 | NM_138426.3 | NP_612435.1 | ||
| BC050291.1 | AAH50291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571615 | Chr.7:8046535 - 8050345 on Build GRCh38 | Loss |
|
| esv2752177 | Chr.7:7689616 - 8138059 on Build GRCh38 | Gain |
|
| esv2761144 | Chr.7:7457249 - 8757480 on Build GRCh38 | Gain+Loss |
|

Genomic Map