Assay Details
Target Gene Details
Entrez Gene ID: | 154791 |
Gene Name: | formation of mitochondrial complex V assembly factor 1 homolog |
Gene Aliases: |
C7orf55, HSPC268 |
Location: |
Chr.7:139340450-139346319 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FMC1 | NR_073058.1 | |||
| NR_073059.1 | ||||
| NR_073060.1 | ||||
| AA393224.1 | ||||
| AA620996.1 | ||||
| BU852633.1 |
Target Gene Details
Entrez Gene ID: | 51631 |
Gene Name: | LUC7 like 2, pre-mRNA splicing factor |
Gene Aliases: |
CGI-59, CGI-74, LUC7B2 |
Location: |
Chr.7:139340359-139423457 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUC7L2 | NM_001244585.1 | NP_001231514.1 | ||
| AK298166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map