Assay Details
Target Gene Details
Entrez Gene ID: | 7982 |
Gene Name: | suppression of tumorigenicity 7 |
Gene Aliases: |
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7 |
Location: |
Chr.7:116953327-117230023 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST7 | NM_018412.3 | NP_060882.2 | ||
| NM_021908.2 | NP_068708.1 | |||
| AF234882.1 | AAF85945.1 | |||
| AF234883.1 | AAF85946.1 | |||
| AK093212.1 | BAC04097.1 | |||
| AK292372.1 | ||||
| AK297510.1 | ||||
| AK297861.1 | ||||
| AK298540.1 | ||||
| AY009152.1 | AAG37720.1 | |||
| AY009153.1 | AAG37721.1 | |||
| BC030954.1 | AAH30954.1 | |||
| BC075855.1 | AAH75855.1 | |||
| KJ534947.1 |
Target Gene Details
Entrez Gene ID: | 93654 |
Gene Name: | ST7 antisense RNA 2 |
Gene Aliases: |
ST7AS2, ST7OT2 |
Location: |
Chr.7:117072072-117145592 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST7-AS2 | NR_109980.1 | |||
| AF400040.1 | ||||
| AF400042.1 | ||||
| CB054673.1 |

Genomic Map