Assay Details
Target Gene Details
Entrez Gene ID: | 154661 |
Gene Name: | RUN domain containing 3B |
Gene Aliases: |
RPIB9, RPIP9 |
Location: |
Chr.7:87627656-87832298 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNDC3B | NM_001134405.1 | NP_001127877.1 | ||
| NM_001134406.1 | NP_001127878.1 | |||
| NM_138290.2 | NP_612147.1 | |||
| XM_005250156.2 | XP_005250213.1 | |||
| XM_005250158.2 | XP_005250215.1 | |||
| XM_011515826.1 | XP_011514128.1 | |||
| XM_011515827.1 | XP_011514129.1 | |||
| XM_011515828.2 | XP_011514130.1 | |||
| XM_011515829.2 | XP_011514131.1 | |||
| XM_017011761.1 | XP_016867250.1 | |||
| XM_017011762.1 | XP_016867251.1 | |||
| XM_017011763.1 | XP_016867252.1 | |||
| XM_017011764.1 | XP_016867253.1 | |||
| XM_017011765.1 | XP_016867254.1 | |||
| XM_017011766.1 | XP_016867255.1 | |||
| AK055233.1 | BAB70882.1 | |||
| AK091536.1 | BAC03686.1 | |||
| AK293607.1 | ||||
| AK294039.1 | ||||
| AY033596.1 | AAK52313.1 | |||
| BC022520.1 | AAH22520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464622 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain |
|
| nsv528078 | Chr.7:87768029 - 87789546 on Build GRCh38 | Loss |
|

Genomic Map