Assay Details
Target Gene Details
Entrez Gene ID: | 5782 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 12 |
Gene Aliases: |
PTP-PEST, PTPG1 |
Location: |
Chr.7:77537160-77640071 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN12 | NM_001131008.1 | NP_001124480.1 | ||
| NM_001131009.1 | NP_001124481.1 | |||
| NM_002835.3 | NP_002826.3 | |||
| XM_005250518.2 | XP_005250575.1 | |||
| XM_006716073.3 | XP_006716136.1 | |||
| XM_011516444.1 | XP_011514746.1 | |||
| XM_017012474.1 | XP_016867963.1 | |||
| AB209524.1 | BAD92761.1 | |||
| AK289573.1 | ||||
| AK296764.1 | ||||
| AK303602.1 | ||||
| AK314236.1 | ||||
| BC050008.1 | AAH50008.2 | |||
| CN395133.1 | ||||
| D13380.1 | BAA02648.1 | |||
| M93425.1 | AAA36529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022292 | Chr.7:77346780 - 77612732 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map