Assay Details
Target Gene Details
Entrez Gene ID: | 1129 |
Gene Name: | cholinergic receptor muscarinic 2 |
Gene Aliases: |
HM2 |
Location: |
Chr.7:136868652-137020255 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRM2 | NM_000739.2 | NP_000730.1 | ||
| NM_001006626.1 | NP_001006627.1 | |||
| NM_001006627.1 | NP_001006628.1 | |||
| NM_001006628.1 | NP_001006629.1 | |||
| NM_001006629.1 | NP_001006630.1 | |||
| NM_001006630.1 | NP_001006631.1 | |||
| NM_001006631.1 | NP_001006632.1 | |||
| NM_001006632.1 | NP_001006633.1 | |||
| AK290236.1 | ||||
| AL832585.1 | ||||
| AY034603.2 | AAO23567.1 | |||
| AY034604.2 | AAO23568.1 | |||
| AY034605.2 | AAO23569.1 | |||
| AY219703.1 | AAO65595.1 | |||
| AY219704.1 | AAO65596.1 | |||
| AY219705.1 | AAO65597.1 | |||
| AY456125.1 | AAR14890.1 | |||
| AY456126.1 | AAR14891.1 | |||
| AY456127.1 | AAR14892.1 |
More Information
Set Membership: |
|

Genomic Map