Assay Details
Target Gene Details
Entrez Gene ID: | 27075 |
Gene Name: | tetraspanin 13 |
Gene Aliases: |
NET-6, NET6, TM4SF13 |
Location: |
Chr.7:16753726-16784536 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN13 | NM_014399.3 | NP_055214.1 | ||
| AF100759.1 | AAD43023.1 | |||
| AF120265.1 | AAD17294.1 | |||
| AK026587.1 | ||||
| AK075475.1 | ||||
| AK093487.1 | ||||
| AK316595.1 | ||||
| AY358995.1 | AAQ89354.1 | |||
| BC033863.1 | AAH33863.1 | |||
| CR542107.1 | CAG46904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677204 | Chr.7:16755037 - 16760872 on Build GRCh38 | Deletion |
|
| esv3612404 | Chr.7:16755027 - 16760868 on Build GRCh38 | Loss |
|
| nsv824019 | Chr.7:16754864 - 16760964 on Build GRCh38 | Loss |
|
| dgv6289n100 | Chr.7:16230707 - 17452994 on Build GRCh38 | Gain |
|
| nsv1028125 | Chr.7:16489535 - 16807601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map