Assay Details
Target Gene Details
Entrez Gene ID: | 5244 |
Gene Name: | ATP binding cassette subfamily B member 4 |
Gene Aliases: |
ABC21, GBD1, ICP3, MDR2, MDR2/3, MDR3, PFIC-3, PGY3 |
Location: |
Chr.7:87399461-87475864 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB4 | XM_011516308.2 | 29 | 5416 | XP_011514610.2 |
| XM_011516309.2 | 29 | 5395 | XP_011514611.2 | |
| XM_011516310.2 | 28 | 5311 | XP_011514612.2 | |
| XM_011516311.2 | 29 | 5287 | XP_011514613.2 | |
| XM_011516312.2 | 28 | 5275 | XP_011514614.2 | |
| XM_011516315.2 | 24 | 4792 | XP_011514617.2 | |
| XM_017012323.1 | 29 | 5403 | XP_016867812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464622 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain |
|
| nsv428177 | Chr.7:87228237 - 87404851 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map