Assay Details
Target Gene Details
Entrez Gene ID: | 107 |
Gene Name: | adenylate cyclase 1 |
Gene Aliases: |
AC1, DFNB44 |
Location: |
Chr.7:45574140-45723116 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY1 | NM_001281768.1 | NP_001268697.1 | ||
| NM_021116.2 | NP_066939.1 | |||
| XM_005249584.3 | XP_005249641.1 | |||
| XM_005249585.2 | XP_005249642.1 | |||
| AB209496.1 | BAD92733.1 | |||
| AF497515.1 | AAM94372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950210 | Chr.7:45572902 - 45578501 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs367978525] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map