Assay Details
Target Gene Details
Entrez Gene ID: | 140545 |
Gene Name: | ring finger protein 32 |
Gene Aliases: |
FKSG33, HSD15, LMBR2 |
Location: |
Chr.7:156640286-156678333 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023469 | Chr.7:156611368 - 156645912 on Build GRCh38 | Gain |
|
| esv3615671 | Chr.7:156639968 - 156644475 on Build GRCh38 | Loss |
|
| nsv831198 | Chr.7:156629947 - 156829662 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map