Assay Details
Target Gene Details
Entrez Gene ID: | 100506497 |
Gene Name: | uncharacterized LOC100506497 |
Gene Aliases: |
- |
Location: |
Chr.7:28979967-29013369 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506497 | NR_038965.1 | |||
| AK124888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612679 | Chr.7:28970964 - 29086873 on Build GRCh38 | Loss |
|
| esv2762660 | Chr.7:28985794 - 28993192 on Build GRCh38 | Loss |
|
| esv2673735 | Chr.7:28984737 - 28994053 on Build GRCh38 | Deletion |
|
| esv3612683 | Chr.7:28984677 - 28994054 on Build GRCh38 | Loss |
|
| esv3612680 | Chr.7:28970982 - 28988583 on Build GRCh38 | Loss |
|

Genomic Map