Assay Details
Target Gene Details
Entrez Gene ID: | 5243 |
Gene Name: | ATP binding cassette subfamily B member 1 |
Gene Aliases: |
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1 |
Location: |
Chr.7:87503863-87713323 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB1 | NM_000927.4 | NP_000918.2 | ||
| AF345623.1 | ||||
| AF345624.1 | ||||
| AF345625.1 | ||||
| AK290159.1 | ||||
| AY425005.1 | AAR91621.1 | |||
| AY425006.1 | AAR91622.1 | |||
| M14758.1 | AAA59575.1 |
Target Gene Details
Entrez Gene ID: | 154661 |
Gene Name: | RUN domain containing 3B |
Gene Aliases: |
RPIB9, RPIP9 |
Location: |
Chr.7:87627656-87832298 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNDC3B | XM_011515829.2 | 1 | 90 | XP_011514131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464622 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map