Assay Details
Target Gene Details
Entrez Gene ID: | 23080 |
Gene Name: | AVL9 cell migration associated |
Gene Aliases: |
KIAA0241 |
Location: |
Chr.7:32495426-32588741 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVL9 | NM_015060.2 | 16 | 4162 | NP_055875.1 |
| XM_005249668.2 | 16 | 4111 | XP_005249725.1 | |
| XM_005249669.2 | 15 | 4057 | XP_005249726.1 | |
| XM_005249670.2 | 16 | 3817 | XP_005249727.1 | |
| XM_011515227.2 | 14 | 3939 | XP_011513529.1 | |
| XM_017011891.1 | 16 | 3805 | XP_016867380.1 | |
| XM_017011892.1 | 16 | 3815 | XP_016867381.1 | |
| BC027724.2 | AAH27724.1 | |||
| D87682.1 | 12 | 3500 | BAA13435.1 |
Target Gene Details
Entrez Gene ID: | 100129460 |
Gene Name: | DPY19L1 pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.7:32580939-32719168 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPY19L1P1 | NR_036680.1 | |||
| AK026768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map