Assay Details
Target Gene Details
Entrez Gene ID: | 23534 |
Gene Name: | transportin 3 |
Gene Aliases: |
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR |
Location: |
Chr.7:128954180-129055173 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNPO3 | NM_001191028.2 | NP_001177957.2 | ||
| NM_012470.3 | NP_036602.1 | |||
| NR_034053.2 | ||||
| XM_011515989.2 | XP_011514291.1 | |||
| AK001263.1 | ||||
| AK022910.1 | ||||
| AK225999.1 | ||||
| BC009221.2 | AAH09221.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670312 | Chr.7:128499519 - 128999613 on Build GRCh38 | Deletion |
|
| esv2735137 | Chr.7:128499440 - 128999664 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map