Assay Details
Target Gene Details
Entrez Gene ID: | 6742 |
Gene Name: | single stranded DNA binding protein 1 |
Gene Aliases: |
Mt-SSB, SOSS-B1, SSBP, mtSSB |
Location: |
Chr.7:141738309-141750488 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP1 | NM_001256510.1 | NP_001243439.1 | ||
| NM_001256511.1 | NP_001243440.1 | |||
| NM_001256512.1 | NP_001243441.1 | |||
| NM_001256513.1 | NP_001243442.1 | |||
| NM_003143.2 | NP_003134.1 | |||
| NR_046269.1 | ||||
| XM_005250048.4 | XP_005250105.1 | |||
| XM_005250049.4 | XP_005250106.1 | |||
| XM_005250050.4 | XP_005250107.1 | |||
| XM_005250051.4 | XP_005250108.1 | |||
| AF277319.1 | AAK69112.1 | |||
| AK294393.1 | ||||
| AK311200.1 | 2 | 1331 | ||
| AK313033.1 | ||||
| BC000895.1 | AAH00895.1 | |||
| BC008402.1 | 2 | 1281 | AAH08402.1 | |
| BC093054.1 | AAH93054.1 | |||
| BF131408.1 | ||||
| BP308841.1 | ||||
| BQ959702.1 | ||||
| BU532904.1 | ||||
| DB464012.1 | ||||
| M94556.1 | AAA36332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34135 | Chr.7:141347654 - 141793098 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map