Assay Details
Target Gene Details
Entrez Gene ID: | 28996 |
Gene Name: | homeodomain interacting protein kinase 2 |
Gene Aliases: |
PRO0593 |
Location: |
Chr.7:139561570-139777894 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIPK2 | NM_001113239.2 | 15 | 7975 | NP_001106710.1 |
| NM_022740.4 | 15 | 8056 | NP_073577.3 | |
| XM_006715935.2 | 16 | 8045 | XP_006715998.1 | |
| XM_011516077.2 | 15 | 8634 | XP_011514379.1 | |
| XM_011516078.2 | 15 | 8632 | XP_011514380.1 | |
| XM_011516079.2 | 15 | 8553 | XP_011514381.1 | |
| XM_011516080.2 | 15 | 8551 | XP_011514382.1 | |
| XM_011516081.2 | 15 | 8031 | XP_011514383.1 | |
| XM_017012069.1 | 15 | 8137 | XP_016867558.1 | |
| AK074291.1 | 1 | 1097 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map