Assay Details
Target Gene Details
Entrez Gene ID: | 148 |
Gene Name: | adrenoceptor alpha 1A |
Gene Aliases: |
ADRA1C, ADRA1L1, ALPHA1AAR |
Location: |
Chr.8:26738113-26870994 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRA1A | XM_006716292.3 | 4 | 4112 | XP_006716355.1 |
| XM_017013094.1 | 5 | 7672 | XP_016868583.1 | |
| XM_017013095.1 | 5 | 7695 | XP_016868584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616704 | Chr.8:26737549 - 26747280 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map