Assay Details
Target Gene Details
Entrez Gene ID: | 103021164 |
Gene Name: | cancer susceptibility candidate 21 (non-protein coding) |
Gene Aliases: |
CARLo-2, LINC01244 |
Location: |
Chr.8:127244637-127392631 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASC21 | NR_117099.1 | |||
| AL709378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3618770 | Chr.8:127255627 - 127259551 on Build GRCh38 | Loss |
|
| esv2669237 | Chr.8:127255604 - 127259560 on Build GRCh38 | Deletion |
|
| nsv831457 | Chr.8:127083126 - 127260605 on Build GRCh38 | Gain |
|

Genomic Map