Assay Details
Target Gene Details
Entrez Gene ID: | 3786 |
Gene Name: | potassium voltage-gated channel subfamily Q member 3 |
Gene Aliases: |
BFNC2, EBN2, KV7.3 |
Location: |
Chr.8:132120858-132480757 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ3 | NM_001204824.1 | 15 | 10072 | NP_001191753.1 |
| NM_004519.3 | 15 | 10621 | NP_004510.1 | |
| XM_005250914.3 | 12 | 9696 | XP_005250971.1 | |
| XM_006716555.3 | 13 | 9882 | XP_006716618.1 | |
| XM_011517026.2 | 15 | 13155 | XP_011515328.1 | |
| XM_017013400.1 | 15 | 20127 | XP_016868889.1 | |
| AK095711.1 | 1 | 2011 | ||
| BX538233.1 | 1 | 4390 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv612325 | Chr.8:131918447 - 132338873 on Build GRCh38 | Gain |
|
| esv3618853 | Chr.8:131375690 - 132136917 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map