Assay Details
Target Gene Details
Entrez Gene ID: | 8658 |
Gene Name: | tankyrase |
Gene Aliases: |
ARTD5, PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5 |
Location: |
Chr.8:9555229-9782346 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNKS | NM_003747.2 | NP_003738.2 | ||
| XM_006716263.3 | XP_006716326.1 | |||
| XM_011543845.2 | XP_011542147.1 | |||
| XM_011543846.2 | XP_011542148.1 | |||
| XM_011543847.2 | XP_011542149.1 | |||
| AB209339.1 | BAD92576.1 | |||
| AF082556.1 | AAC79841.1 | |||
| AF082557.1 | AAC79842.1 | |||
| AK301798.1 | ||||
| AK302031.1 | ||||
| AK302534.1 | ||||
| BC098394.1 | AAH98394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3769n106 | Chr.8:9594639 - 9596990 on Build GRCh38 | Deletion |
|
| esv3891335 | Chr.8:9587919 - 9596465 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73535841] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map