Assay Details
Target Gene Details
Entrez Gene ID: | 100506365 |
Gene Name: | OTUD6B antisense RNA 1 (head to head) |
Gene Aliases: |
GS1-251I9.4 |
Location: |
Chr.8:91059906-91070189 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTUD6B-AS1 | NR_110438.1 | |||
| BM715864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831390 | Chr.8:91024783 - 91186377 on Build GRCh38 | Gain |
|
| esv2759629 | Chr.8:90962271 - 91261496 on Build GRCh38 | Loss |
|

Genomic Map