Assay Details
Target Gene Details
Entrez Gene ID: | 5862 |
Gene Name: | RAB2A, member RAS oncogene family |
Gene Aliases: |
LHX, RAB2 |
Location: |
Chr.8:60516910-60623644 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB2A | NM_001242644.1 | NP_001229573.1 | ||
| NM_002865.2 | NP_002856.1 | |||
| AF498930.1 | AAM21078.1 | |||
| AK297576.1 | ||||
| AK312344.1 | ||||
| BC008929.2 | AAH08929.1 | |||
| BT019695.1 | AAV38501.1 | |||
| DA648682.1 | ||||
| M28213.1 | AAA60241.1 | |||
| X12953.1 | CAA31411.1 | |||
| X13899.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73259409] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map