Assay Details
Target Gene Details
Entrez Gene ID: | 7534 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta |
Gene Aliases: |
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD |
Location: |
Chr.8:100918576-100954068 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv971342 | Chr.8:100923589 - 100924905 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map