Assay Details
Target Gene Details
Entrez Gene ID: | 9166 |
Gene Name: | estrogen receptor binding site associated, antigen, 9 |
Gene Aliases: |
EB9, PDAF |
Location: |
Chr.8:109539700-109565996 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBAG9 | NM_001278938.1 | NP_001265867.1 | ||
| NM_004215.4 | NP_004206.1 | |||
| NM_198120.2 | NP_936056.1 | |||
| XM_017013960.1 | XP_016869449.1 | |||
| AB007619.1 | BAA22572.1 | |||
| AF006265.1 | AAB61617.1 | |||
| AK290651.1 | ||||
| AK308950.1 | ||||
| AL515533.3 | ||||
| AY515724.1 | AAR97974.1 | |||
| AY653072.1 | AAU85838.1 | |||
| BC005249.1 | AAH05249.1 | |||
| BC017729.1 | AAH17729.1 | |||
| BC022506.1 | AAH22506.1 | |||
| BG616704.1 | ||||
| BG707115.1 | ||||
| CD108096.1 | ||||
| CR456984.1 | CAG33265.1 |
More Information
Set Membership: |
|

Genomic Map