Assay Details
Target Gene Details
Entrez Gene ID: | 101410533 |
Gene Name: | YTHDF3 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.8:63167725-63168442 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YTHDF3-AS1 | NR_102684.1 | 1 | 132 | |
| BM681776.1 | 1 | 442 | ||
| BQ631682.1 | 1 | 126 |

Genomic Map