Assay Details
Target Gene Details
Entrez Gene ID: | 104054148 |
Gene Name: | uncharacterized LOC104054148 |
Gene Aliases: |
- |
Location: |
Chr.8:101984432-101994850 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC104054148 | NR_126152.1 | 4 | 2095 | |
| AK093137.1 | 4 | 2099 |
Target Gene Details
Entrez Gene ID: | 83988 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
- |
Location: |
Chr.8:101686542-102124907 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCALD | NM_001040624.1 | NP_001035714.1 | ||
| NM_001040625.1 | NP_001035715.1 | |||
| NM_001040626.1 | NP_001035716.1 | |||
| NM_001040627.1 | NP_001035717.1 | |||
| NM_001040628.1 | NP_001035718.1 | |||
| NM_001040629.1 | NP_001035719.1 | |||
| NM_001040630.1 | NP_001035720.1 | |||
| XM_011517333.2 | XP_011515635.1 | |||
| XM_017013900.1 | XP_016869389.1 | |||
| XM_017013901.1 | XP_016869390.1 | |||
| AF251061.1 | AAK34951.1 | |||
| AK096515.1 | ||||
| BC036098.1 | AAH36098.1 | |||
| BC063428.1 | AAH63428.1 | |||
| DA185335.1 | ||||
| DA215759.1 | ||||
| DA417037.1 | ||||
| DA776593.1 | ||||
| DA779293.1 | ||||
| DB284042.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3618213 | Chr.8:101986638 - 101998483 on Build GRCh38 | Loss |
|
| esv3618214 | Chr.8:101987327 - 102007840 on Build GRCh38 | Loss |
|
| nsv831411 | Chr.8:101925763 - 102095912 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map