Assay Details
Target Gene Details
Entrez Gene ID: | 286135 |
Gene Name: | family with sequence similarity 183 member C, pseudogene |
Gene Aliases: |
- |
Location: |
Chr.8:29921513-29953609 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM183CP | NR_024473.1 | |||
| AL833525.1 |

Genomic Map