Assay Details
Target Gene Details
Entrez Gene ID: | 11075 |
Gene Name: | stathmin 2 |
Gene Aliases: |
SCG10, SCGN10 |
Location: |
Chr.8:79610814-79666175 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STMN2 | NM_001199214.1 | NP_001186143.1 | ||
| NM_007029.3 | NP_008960.2 | |||
| XM_005251142.2 | XP_005251199.1 | |||
| AK292737.1 | ||||
| AK297485.1 | ||||
| AK297594.1 | ||||
| AK299500.1 | ||||
| AK308557.1 | ||||
| AK309130.1 | ||||
| BC006302.2 | AAH06302.1 | |||
| BQ069488.1 | ||||
| D50375.1 | BAA23326.1 | |||
| DC319376.1 | ||||
| S82024.1 | AAB36428.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78260664] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map