Assay Details
Target Gene Details
Entrez Gene ID: | 103021164 |
Gene Name: | cancer susceptibility candidate 21 (non-protein coding) |
Gene Aliases: |
CARLo-2, LINC01244 |
Location: |
Chr.8:127244637-127392631 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASC21 | NR_117099.1 | |||
| AL709378.1 |

Genomic Map