Assay Details
Target Gene Details
Entrez Gene ID: | 9312 |
Gene Name: | potassium voltage-gated channel subfamily B member 2 |
Gene Aliases: |
KV2.2 |
Location: |
Chr.8:72537391-72945890 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNB2 | NM_004770.2 | NP_004761.2 | ||
| AF338730.1 | AAK16585.1 | |||
| AK311263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025110 | Chr.8:72091240 - 72818631 on Build GRCh38 | Gain |
|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115275719] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map