Assay Details
Target Gene Details
Entrez Gene ID: | 3174 |
Gene Name: | hepatocyte nuclear factor 4 gamma |
Gene Aliases: |
NR2A2, NR2A3 |
Location: |
Chr.8:75407567-75566834 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNF4G | XM_011517516.2 | XP_011515818.1 | ||
| XM_017013373.1 | XP_016868862.1 | |||
| XM_017013374.1 | XP_016868863.1 | |||
| XM_017013375.1 | XP_016868864.1 | |||
| BC054009.1 | AAH54009.1 | |||
| Z49826.2 | CAA89990.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611561 | Chr.8:75288007 - 75914413 on Build GRCh38 | Gain |
|
| esv3576590 | Chr.8:74924274 - 75416508 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map