Assay Details
Target Gene Details
Entrez Gene ID: | 8754 |
Gene Name: | ADAM metallopeptidase domain 9 |
Gene Aliases: |
CORD9, MCMP, MDC9, Mltng |
Location: |
Chr.8:38996771-39105261 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM9 | NM_003816.2 | NP_003807.1 | ||
| NR_027638.1 | ||||
| NR_027639.1 | ||||
| NR_027878.1 | ||||
| XM_011544682.2 | XP_011542984.1 | |||
| AF495383.1 | AAM49575.1 | |||
| BC126406.1 | ||||
| BC143923.1 | ||||
| U41766.1 | AAC50403.1 |
More Information
Set Membership: |
|

Genomic Map