Assay Details
Target Gene Details
Entrez Gene ID: | 157657 |
Gene Name: | chromosome 8 open reading frame 37 |
Gene Aliases: |
BBS21, CORD16, RP64, smalltalk |
Location: |
Chr.8:95244913-95270602 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf37 | NM_177965.3 | NP_808880.1 | ||
| XM_005250799.3 | XP_005250856.2 | |||
| AK055162.1 | BAB70866.1 | |||
| DA473498.1 | ||||
| DB205618.1 | ||||
| GQ240139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525440 | Chr.8:95242118 - 95264064 on Build GRCh38 | Loss |
|
| nsv6315 | Chr.8:95204237 - 95249443 on Build GRCh38 | Deletion |
|
| nsv819165 | Chr.8:95247287 - 95248635 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73695435] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map