Assay Details
Target Gene Details
Entrez Gene ID: | 51115 |
Gene Name: | regulator of microtubule dynamics 1 |
Gene Aliases: |
CGI-90, FAM82B, RMD-1, RMD1 |
Location: |
Chr.8:86467116-86514348 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN1 | XM_011517090.2 | XP_011515392.1 | ||
| XM_011517095.2 | XP_011515397.1 | |||
| XM_017013518.1 | XP_016869007.1 | |||
| XM_017013525.1 | XP_016869014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023510 | Chr.8:86416772 - 86862720 on Build GRCh38 | Loss |
|

Genomic Map