Assay Details
Target Gene Details
Entrez Gene ID: | 90362 |
Gene Name: | family with sequence similarity 110 member B |
Gene Aliases: |
C8orf72 |
Location: |
Chr.8:57994444-58149718 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM110B | NM_147189.2 | NP_671722.1 | ||
| XM_005251324.2 | XP_005251381.1 | |||
| AK092221.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831324 | Chr.8:57931592 - 58132750 on Build GRCh38 | Loss |
|

Genomic Map