Assay Details
Target Gene Details
Entrez Gene ID: | 22898 |
Gene Name: | DENN domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.8:141128618-141195804 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND3 | NM_014957.2 | NP_055772.2 | ||
| XM_005250838.4 | XP_005250895.2 | |||
| XM_005250839.2 | XP_005250896.2 | |||
| XM_005250840.4 | XP_005250897.2 | |||
| XM_005250841.3 | XP_005250898.2 | |||
| XM_005250842.4 | XP_005250899.1 | |||
| XM_011516933.1 | XP_011515235.1 | |||
| XM_011516934.2 | XP_011515236.1 | |||
| XM_011516937.1 | XP_011515239.1 | |||
| XM_011516941.2 | XP_011515243.1 | |||
| XM_017013241.1 | XP_016868730.1 | |||
| XM_017013242.1 | XP_016868731.1 | |||
| AK058001.1 | BAB71630.1 | |||
| AL544320.3 | ||||
| CB989310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2737756 | Chr.8:140419230 - 141917755 on Build GRCh38 | Deletion |
|
| nsv612568 | Chr.8:140534649 - 141326581 on Build GRCh38 | Gain |
|
| nsv831483 | Chr.8:141123462 - 141290370 on Build GRCh38 | Gain |
|
| dgv1426e214 | Chr.8:141120386 - 141147214 on Build GRCh38 | Loss |
|
| nsv509284 | Chr.8:141010114 - 141129733 on Build GRCh38 | Insertion |
|
| nsv1126125 | Chr.8:140419236 - 141917668 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map