Assay Details
Target Gene Details
Entrez Gene ID: | 3574 |
Gene Name: | interleukin 7 |
Gene Aliases: |
IL-7 |
Location: |
Chr.8:78675870-78805523 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL7 | XM_011517523.2 | XP_011515825.1 |
Target Gene Details
Entrez Gene ID: | 51101 |
Gene Name: | zinc finger C2HC-type containing 1A |
Gene Aliases: |
C8orf70, CGI-62, FAM164A |
Location: |
Chr.8:78666047-78719765 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC2HC1A | NM_016010.2 | NP_057094.2 | ||
| XM_005251256.1 | XP_005251313.1 | |||
| XM_006716454.2 | XP_006716517.1 | |||
| XM_011517539.2 | XP_011515841.1 | |||
| AF151820.1 | AAD34057.1 | |||
| AK024711.1 | ||||
| AK313723.1 | ||||
| BC009074.1 | AAH09074.1 | |||
| DA137885.1 |
More Information
Set Membership: |
|

Genomic Map