Assay Details
Target Gene Details
Entrez Gene ID: | 101928525 |
Gene Name: | uncharacterized LOC101928525 |
Gene Aliases: |
- |
Location: |
Chr.9:135503273-135506447 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928525 | NR_121579.1 | 1 | 416 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615842 | Chr.9:135113632 - 135571320 on Build GRCh38 | Gain |
|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| esv3573422 | Chr.9:135497894 - 135508556 on Build GRCh38 | Loss |
|
| nsv466656 | Chr.9:135491367 - 135577889 on Build GRCh38 | Gain |
|

Genomic Map